Learn More
Medchemexpress LLC HY-117937 5mg Medchemexpress, BC-1215 CAS:1507370-20-8 Purity:>98%
SDP
Catalog No. 501969497
Encompass_Preferred
Medchemexpress, HY-117937 5mg BC-1215 CAS:1507370-20-8 BC-1215 is an inhibitor of F-box protein 3 (FBXO3, a ubiquitin E3 ligase component, IC50=0.9 μg/mL for IL-1β release). BC-1215 decreases Fbxo3-Fbxl2 interaction and prevents SCFFbxo3 catalyzed Fbxl2 ubiquitination. BC-1215 inhibits the Fbxo3-TRAF activation pathway by destabilizing TRAF1–TRAF6. BC-1215, interacts with ApaG to profoundly inhibit secretion of a broad spectrum of TH1 panel cytokines from human PBMC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9497 Supplier Medchemexpress LLC Supplier No. HY1179375MG
Medchemexpress, HY-117937 5mg BC-1215 CAS:1507370-20-8 BC-1215 is an inhibitor of F-box protein 3 (FBXO3, a ubiquitin E3 ligase component, IC50=0.9 μg/mL for IL-1β release). BC-1215 decreases Fbxo3-Fbxl2 interaction and prevents SCFFbxo3 catalyzed Fbxl2 ubiquitination. BC-1215 inhibits the Fbxo3-TRAF activation pathway by destabilizing TRAF1–TRAF6. BC-1215, interacts with ApaG to profoundly inhibit secretion of a broad spectrum of TH1 panel cytokines from human PBMC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.