Learn More
TARGETMOL CHEMICALS INC BC-1471 5MG
SDP
Catalog No. 502579061
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BC-1471 is a STAM-binding protein (STAMBP) deubiquitinase inhibitor. BC-1471 inhibits inflammasome activity of NALP7 (NACHT LRR and PYD domains-containing protein 7) [1]. purity: 98%
Catalog No. 50-257-9061 Supplier TARGETMOL CHEMICALS INC Supplier No. T85535MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BC-1471 is a STAM-binding protein (STAMBP) deubiquitinase inhibitor. BC-1471 inhibits inflammasome activity of NALP7 (NACHT LRR and PYD domains-containing protein 7) [1]. purity: 98%