Learn More
TARGETMOL CHEMICALS INC BC-1485 1MG
SDP
Catalog No. 502877835
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. BC-1485 is a small molecule inhibitor of FIEL1 (Fibrosis Inducing E3 Ligase 1) that inhibits the degradation of PIAS4 and ameliorates fibrosis in a mouse model. Purity 98.61%
Catalog No. 50-287-7835 Supplier TARGETMOL CHEMICALS INC Supplier No. T699771MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. BC-1485 is a small molecule inhibitor of FIEL1 (Fibrosis Inducing E3 Ligase 1) that inhibits the degradation of PIAS4 and ameliorates fibrosis in a mouse model. Purity 98.61%