Learn More
Selleck Chemical LLC BC1618-5mg
SDP
Catalog No. 502389594
Encompass_Preferred
BC1618 is an orally active Fbxo48 inhibitor that stimulates adenosine monophosphate (AMP)-activated protein kinase (Ampk)-dependent signaling. BC1618 promotes mitochondrial fission, facilitates autophagy and improves hepatic insulin sensitivity.
Catalog No. 50-238-9594 Supplier Selleck Chemical LLC Supplier No. S98605MG
BC1618 is an orally active Fbxo48 inhibitor that stimulates adenosine monophosphate (AMP)-activated protein kinase (Ampk)-dependent signaling. BC1618 promotes mitochondrial fission, facilitates autophagy and improves hepatic insulin sensitivity.