Learn More
Aviva Systems Biology Corp ZNF835 Antibody - N-terminal region (ARP50335_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50335_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BC37295_3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BC37295_3. Alternate names: BC37295_3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60318