Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bca-1/CXCL13 protein, human (His) | >95.0% | 100UG
SDP

Catalog No. 5000173955
Encompass_Preferred

Recombinant human BCA-1/CXCL13 (His-tagged) is a CXC chemokine that selectively attracts B lymphocytes via CXCR5. The lyophilized protein is supplied for research use in in vitro assays, chemotaxis, and biochemical characterization.

  • His-tagged recombinant protein, amino acids V23-P109.
  • Expressed in E. coli and lyophilized for stability.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Formulated in PBS with 10% trehalose and 0.22 μm filtered.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.

Catalog No. 50-001-73955 Supplier Medchemexpress LLC Supplier No. HYP704362100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human BCA-1/CXCL13 (His-tagged) is a CXC chemokine that selectively attracts B lymphocytes via CXCR5. The lyophilized protein is supplied for research use in in vitro assays, chemotaxis, and biochemical characterization.

  • His-tagged recombinant protein, amino acids V23-P109.
  • Expressed in E. coli and lyophilized for stability.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Formulated in PBS with 10% trehalose and 0.22 μm filtered.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.