Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BCA-1/CXCL13 Human 1mg
SDP

Catalog No. 5000196529
Encompass_Preferred

Recombinant human BCA-1 (CXCL13) is a tag-free chemokine corresponding to residues V23-P109, supplied lyophilized for biochemical and cell-based assays. Expressed in E. coli and purified to >95% purity by reducing SDS-PAGE, this ~11 kDa protein is suitable for chemotaxis, binding, and structural studies when reconstituted and stored according to the product guidelines.

  • Tag free, matching human sequence (V23-P109).
  • Expressed in E. coli for efficient production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~11 kDa for easy characterization.
  • Lyophilized formulation for improved stability and handling.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and freeze for long-term storage.

Catalog No. 50-001-96529 Supplier Medchemexpress LLC Supplier No. HYP71311MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human BCA-1 (CXCL13) is a tag-free chemokine corresponding to residues V23-P109, supplied lyophilized for biochemical and cell-based assays. Expressed in E. coli and purified to >95% purity by reducing SDS-PAGE, this ~11 kDa protein is suitable for chemotaxis, binding, and structural studies when reconstituted and stored according to the product guidelines.

  • Tag free, matching human sequence (V23-P109).
  • Expressed in E. coli for efficient production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~11 kDa for easy characterization.
  • Lyophilized formulation for improved stability and handling.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and freeze for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.