Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BCA-1/CXCL13 protein, rat (His-GST) | >90.0% | 20UG
SDP

Catalog No. 5000176468
Encompass_Preferred

Recombinant rat BCA-1 (CXCL13) is a CXC chemokine produced with an N-terminal His-GST tag in Escherichia coli and supplied lyophilized for research use. The protein is suitable for receptor binding studies, chemotaxis assays, and antibody generation, and is provided with characterized purity, molecular weight, and storage recommendations.

  • Species: rat
  • Tag: N-terminal His-GST
  • Expression system: Escherichia coli
  • Purity: >90.0%
  • Predicted molecular weight: 36.8 kDa
  • Formulation: lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0
  • Storage: store at -20°C; stable for 2 years from receipt
  • Available sizes: 20 μg, 50 μg, 100 μg; larger sizes on request

Catalog No. 50-001-76468 Supplier Medchemexpress LLC Supplier No. HYP70054720UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat BCA-1 (CXCL13) is a CXC chemokine produced with an N-terminal His-GST tag in Escherichia coli and supplied lyophilized for research use. The protein is suitable for receptor binding studies, chemotaxis assays, and antibody generation, and is provided with characterized purity, molecular weight, and storage recommendations.

  • Species: rat
  • Tag: N-terminal His-GST
  • Expression system: Escherichia coli
  • Purity: >90.0%
  • Predicted molecular weight: 36.8 kDa
  • Formulation: lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0
  • Storage: store at -20°C; stable for 2 years from receipt
  • Available sizes: 20 μg, 50 μg, 100 μg; larger sizes on request

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.