Learn More
Cell Signaling Technology BCA Protein Assay Kit, 960 Assays, 0.125 to 2 mg/mL Dynamic Range, 1 Kit
SDP
Catalog No. 502050890
Encompass
The BCA Protein Assay Kit provides a method to measure protein concentration of lysates or homogenates in a microplate format. It is based on the formation of a purple complex between bicinchoninic acid and cuprous ions in an alkaline environment, where color intensity correlates to protein concentration.
  • Dynamic range of 0.125 to 2 mg/mL
  • Compatible with common lysis buffers including Cell Lysis Buffer (10X), RIPA Buffer (10X), and Sandwich ELISA Lysis Buffer (1X)
  • Includes compatibility reagent to decrease interference from reducing agents, chelators, and detergents
  • Suitable for 960 assays
  • Store at room temperature
Catalog No. 50-205-0890 Supplier Cell Signaling Technology Supplier No. 7780S
The BCA Protein Assay Kit provides a method to measure protein concentration of lysates or homogenates in a microplate format. It is based on the formation of a purple complex between bicinchoninic acid and cuprous ions in an alkaline environment, where color intensity correlates to protein concentration.
  • Dynamic range of 0.125 to 2 mg/mL
  • Compatible with common lysis buffers including Cell Lysis Buffer (10X), RIPA Buffer (10X), and Sandwich ELISA Lysis Buffer (1X)
  • Includes compatibility reagent to decrease interference from reducing agents, chelators, and detergents
  • Suitable for 960 assays
  • Store at room temperature