Learn More
Cayman Chemical BCA Reagent 1, 100 mL
SDP
Catalog No. 502894135
Encompass_Preferred
BCA reagent 1 has been tested and formulated to work exclusively with Cayman’s Protein Determination (BCA) Kit (Item No. 701780).
Catalog No. 50-289-4135 Supplier Cayman Chemical Supplier No. 701786100 ML
BCA reagent 1 has been tested and formulated to work exclusively with Cayman’s Protein Determination (BCA) Kit (Item No. 701780).