Learn More
Cayman Chemical BCA Reagent 2, 5 mL
SDP
Catalog No. 502892814
Encompass_Preferred
BCA reagent 2 has been tested and formulated to work exclusively with Cayman’s Protein Determination (BCA) Kit (Item No. 701780).
Catalog No. 50-289-2814 Supplier Cayman Chemical Supplier No. 7017875 ML
BCA reagent 2 has been tested and formulated to work exclusively with Cayman’s Protein Determination (BCA) Kit (Item No. 701780).