Learn More
Aviva Systems Biology Corp BCAM Antibody - C-terminal region (ARP60228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCAM. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BCAM. Alternate names: AU, CD239, LU, MSK19ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-286-981