Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BCAM ELISA Kit

Catalog No. EHBCAMX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHBCAMX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHBCAMX10 Supplier Invitrogen™ Supplier No. EHBCAMX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

The Human Basal Cell Adhesion Molecule (Hu BCAM) ELISA quantitates Hu BCAM in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu BCAM. Principle of the method The Human BCAM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P50895
Assay Range 40.96-10000 pg/mL
Assay Sensitivity 40 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1200005K12Rik; antigen identified by monoclonal antibody F8; AU; Auberger B antigen; basal cell adhesion molecule; basal cell adhesion molecule (Lu and Au blood groups); basal cell adhesion molecule (Lutheran blood group); basal cell adhesion molecule Lutheran blood group; BCAM; B-CAM; B-CAM cell surface glycoprotein; B-cell adhesion molecule; CD239; F8/G253 antigen; glycoprotein 95kDa; Gplu; LU; lutheran antigen; Lutheran blood group; Lutheran blood group (Auberger b antigen included); Lutheran blood group glycoprotein; Lutheran blood group variant LUGA; lutheran glycoprotein; MSK191200005K12Rik; antigen identified by monoclonal antibody F8; AU; Auberger B antigen; basal cell adhesion molecule; basal cell adhesion molecule (Lu and Au blood groups); basal cell adhesion molecule (Lutheran blood group); basal cell adhesion molecule Lutheran blood group; BCAM; B-CAM; B-CAM cell surface glycoprotein; B-cell adhesion molecule; CD239; F8/G253 antigen; glycoprotein 95kDa; Gplu; LU; lutheran antigen; Lutheran blood group; Lutheran blood group (Auberger b antigen included); Lutheran blood group glycoprotein; Lutheran blood group variant LUGA; lutheran glycoprotein; MSK19; AU; CD239; LU; MSK19
Gene ID (Entrez) 4059
Gene Symbol BCAM
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target BCAM
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.