Learn More
Aviva Systems Biology Corp BCAN Antibody - middle region (ARP57797_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57797_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCAN. It was validated on Western Blot. BCAN,brevican,BEHAB; CSPG7; MGC13038; BCAN,brevican core protein,brevican core protein,brevican proteoglycan,chondroitin sulfate proteoglycan 7,chondroitin sulfate proteoglycan BEHAB,brain-enriched hyaluronan-binding protein,chondroitin sulfate proteoglycan BEHAB/brevicanThe immunogen is a synthetic peptide directed towards the middle region of human BCAN. Alternate names: BEHAB, CSPG7, MGC13038ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-381