Learn More
Aviva Systems Biology Corp BCAP29 Antibody - middle region (ARP44557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCAP29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BCAP29. Alternate names: BAP29, DKFZp686M2086ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58842