Learn More
Medchemexpress LLC HY-141668 5mg , BCAT-IN-1 CAS:1875078-61-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1416685MG
Encompass_Preferred
HY-141668 5mg BCAT-IN-1 CAS: 1875078-61-7 BCAT-IN-1 is a potent, selective and orally active inhibitor of BCATm, with a pIC50 of 7.3. BCAT-IN-1 shows 100-fold selectivity for BCATm over BCATc (pIC50=5.4). BCAT-IN-1 can be used for the research of metabolic diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1841