Learn More
Aviva Systems Biology Corp BCAT1 Antibody - N-terminal region (ARP46132_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46132_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BCAT1. Alternate names: BCT1, DKFZp686E12175, ECA39, MECA39, PNAS-121, PP18, BCATC, PNAS121ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Sheep
Catalog No. 50-014-59243