Learn More
Aviva Systems Biology Corp Bcat1 Antibody - C-terminal region (ARP46133_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46133_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bcat1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: BCATc, Eca39ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-243