Learn More
ABclonal Technology BCAT2 Rabbit mAb
SDP
Catalog No. 5000005572
Encompass_Preferred
This gene encodes a branched chain aminotransferase found in mitochondria The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine isoleucine and valine Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-05572 Supplier ABclonal Technology Supplier No. A2379320UL
This gene encodes a branched chain aminotransferase found in mitochondria The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine isoleucine and valine Multiple transcript variants encoding different isoforms have been found for this gene