Learn More
Medchemexpress LLC BCATc Inhibitor 2 10mg | 406191-34-2 | 418.77 g/mol | C16H10ClF3N2O4S | 10 MG
SDP
Catalog No. 5000227941
Encompass_Preferred
BCATc Inhibitor 2 is a selective inhibitor of cytosolic branched-chain aminotransferase (BCATc/BCAT1) used as a research tool in studies of branched-chain amino acid metabolism and neurodegenerative disease mechanisms. Reported IC50 values include 0.2 μM (rat cytosolic rBCATc), 0.8 μM (human cytosolic hBCATc), and 3.0 μM (rat mitochondrial rBCATm). The compound is supplied in high-purity solid and DMSO solution formats for laboratory research.
  • Selective inhibitor of cytosolic branched-chain aminotransferase (BCATc).
  • Reported IC50s: 0.2 μM (rBCATc), 0.8 μM (hBCATc), 3.0 μM (rBCATm).
  • High purity (99.28%).
  • Molecular formula C16H10ClF3N2O4S; molecular weight 418.77 g/mol.
  • Suitable for in vitro biochemical and cell-based assays.
Catalog No. 50-002-27941 Supplier Medchemexpress LLC Supplier No. HY11604410MG
BCATc Inhibitor 2 is a selective inhibitor of cytosolic branched-chain aminotransferase (BCATc/BCAT1) used as a research tool in studies of branched-chain amino acid metabolism and neurodegenerative disease mechanisms. Reported IC50 values include 0.2 μM (rat cytosolic rBCATc), 0.8 μM (human cytosolic hBCATc), and 3.0 μM (rat mitochondrial rBCATm). The compound is supplied in high-purity solid and DMSO solution formats for laboratory research.
  • Selective inhibitor of cytosolic branched-chain aminotransferase (BCATc).
  • Reported IC50s: 0.2 μM (rBCATc), 0.8 μM (hBCATc), 3.0 μM (rBCATm).
  • High purity (99.28%).
  • Molecular formula C16H10ClF3N2O4S; molecular weight 418.77 g/mol.
  • Suitable for in vitro biochemical and cell-based assays.