Learn More
Cayman Chemical BCATc Inhibitor 2, 406191-34-2, 1 mg
SDP
Supplier:  Cayman Chemical 90020021
Encompass_Preferred
Molecular Formula C16H10ClF3N2O4S, Purity ≥98%, Formula Weight 418.8, Synonyms: 5-chloro-2-benzofurancarboxylic acid 2-[[2-(trifluoromethyl)phenyl]sulfonyl]hydrazide, Cytosolic Branched-Chain Amino Acid Transferase Inhibitor 2; A sulfonyl hydrazide that inhibits BCATc (IC50s = 0.81 and 0.2 M in human and rat, respectively) with 15-fold selectivity over BCATm; blocks calcium influx into neurons (IC50 = 4.8 M) following inhibition of glutamate uptake and demonstrates neuroprotective efficacy in vivo.
Catalog No. NC1141883