Learn More
Medchemexpress LLC BCATc Inhibitor 2 25mg | 406191-34-2 | 25 MG
SDP
Catalog No. 5000229252
Encompass_Preferred
BCATc Inhibitor 2 is a selective small-molecule inhibitor of cytosolic branched-chain aminotransferase (BCATc) used in biochemical and neurodegeneration research. It is supplied as a solid and shows greater potency for cytosolic BCAT isoforms than for mitochondrial isoforms.
  • Selective BCAT inhibitor with greater potency for BCATc than BCATm
  • Reported IC50: 0.2 μM (rat BCATc), 0.8 μM (human BCATc), 3.0 μM (rat BCATm)
  • Chemical formula C16H10ClF3N2O4S; molecular weight ~418.8 g/mol
  • White to off-white solid suitable for in vitro research
  • Certificate of analysis indicates typical purity around 98%
Catalog No. 50-002-29252 Supplier Medchemexpress LLC Supplier No. HY11604425MG
BCATc Inhibitor 2 is a selective small-molecule inhibitor of cytosolic branched-chain aminotransferase (BCATc) used in biochemical and neurodegeneration research. It is supplied as a solid and shows greater potency for cytosolic BCAT isoforms than for mitochondrial isoforms.
  • Selective BCAT inhibitor with greater potency for BCATc than BCATm
  • Reported IC50: 0.2 μM (rat BCATc), 0.8 μM (human BCATc), 3.0 μM (rat BCATm)
  • Chemical formula C16H10ClF3N2O4S; molecular weight ~418.8 g/mol
  • White to off-white solid suitable for in vitro research
  • Certificate of analysis indicates typical purity around 98%