Learn More
Medchemexpress LLC BCATc Inhibitor 2 50mg | 406191-34-2 | 418.77 g/mol | C16H10ClF3N2O4S | 50 MG
SDP
Catalog No. 5000227658
Encompass_Preferred
BCATc Inhibitor 2 is a selective small-molecule inhibitor of the cytosolic branched-chain aminotransferase (BCATc/BCAT1), used in laboratory research to probe branched-chain amino acid metabolism and neurodegenerative disease mechanisms. The compound is supplied as a solid and reported to have high purity.
  • Selective BCATc inhibitor with reported IC50s of 0.2 μM (rat rBCATc) and 0.8 μM (human hBCATc)
  • Shows lower potency on mitochondrial BCATm (reported ~3.0 μM)
  • High reported purity (99.28%) suitable for research applications
  • Solid, white to off-white physical form for standard laboratory handling
  • Molecular weight 418.77 g/mol and formula C16H10ClF3N2O4S
Catalog No. 50-002-27658 Supplier Medchemexpress LLC Supplier No. HY11604450MG
BCATc Inhibitor 2 is a selective small-molecule inhibitor of the cytosolic branched-chain aminotransferase (BCATc/BCAT1), used in laboratory research to probe branched-chain amino acid metabolism and neurodegenerative disease mechanisms. The compound is supplied as a solid and reported to have high purity.
  • Selective BCATc inhibitor with reported IC50s of 0.2 μM (rat rBCATc) and 0.8 μM (human hBCATc)
  • Shows lower potency on mitochondrial BCATm (reported ~3.0 μM)
  • High reported purity (99.28%) suitable for research applications
  • Solid, white to off-white physical form for standard laboratory handling
  • Molecular weight 418.77 g/mol and formula C16H10ClF3N2O4S