Learn More
New England Biolabs, Inc. BccI – 1000 units
SDP
Catalog No. 50812414
Encompass
A restriction endonuclease that recognizes the sequence CCATCNNNN^N_.
Catalog No. 50-812-414 Supplier New England Biolabs, Inc. Supplier No. R0704S
A restriction endonuclease that recognizes the sequence CCATCNNNN^N_.