Learn More
Medchemexpress LLC HY-52112 100mg Medchemexpress, BCDA CAS:3252-36-6 Purity:>98%
SDP
Catalog No. 502082262
Encompass_Preferred
Medchemexpress, HY-52112 100mg BCDA CAS:3252-36-6 BCDA (5-bromo-4-chloroindoxyl acetate) is a chromogenic substrate of esterase used to potently detect the activity of esterase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2262 Supplier Medchemexpress LLC Supplier No. HY52112100MG
Medchemexpress, HY-52112 100mg BCDA CAS:3252-36-6 BCDA (5-bromo-4-chloroindoxyl acetate) is a chromogenic substrate of esterase used to potently detect the activity of esterase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.