Learn More
New England Biolabs, Inc. BceAI – 250 units
SDP
Catalog No. 50812363
Encompass
A restriction endonuclease that recognizes the sequence ACGGCNNNNNNNNNNNN^NN_.
Catalog No. 50-812-363 Supplier New England Biolabs, Inc. Supplier No. R0623L
A restriction endonuclease that recognizes the sequence ACGGCNNNNNNNNNNNN^NN_.