Learn More
New England Biolabs, Inc. BceAI – 50 units
SDP
Catalog No. 50812364
Encompass
A restriction endonuclease that recognizes the sequence ACGGCNNNNNNNNNNNN^NN_.
Catalog No. 50-812-364 Supplier New England Biolabs, Inc. Supplier No. R0623S
A restriction endonuclease that recognizes the sequence ACGGCNNNNNNNNNNNN^NN_.