Learn More
AdipoGen BCECF acid,Chemical. CAS: 85138-49-4
SDP
Supplier:  AdipoGen CDXC0002M001
Encompass_Preferred
BCECF acid, AdipoGen Life Sciences. Chemical. CAS: 85138-49-4. Formula: C27H20O11. MW: 520.4. Synthetic. Fluorescent indicator for measurement of cytoplasmic pH in lymphocytes.
Catalog No. 50-101-6848