Learn More
Biotium BCECF, AM Ester, 10x100UG
Catalog No. 501964702
Encompass_Preferred
BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells. Properties: White solid soluble in DMSO and DMF; Store at -20°C and protect from light, especially in solution; MW: approx. 615; [117464-70-7]
Catalog No. 50-196-4702 Supplier Biotium Supplier No. 51011
BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells. Properties: White solid soluble in DMSO and DMF; Store at -20°C and protect from light, especially in solution; MW: approx. 615; [117464-70-7]