Learn More
Sigma Aldrich Fine Chemicals Biosciences BCG Albumin Assay Kit suff
Catalog No. 502904889
Encompass
Albumin is the most abundant plasma protein in humans. It accounts for 60% of the total serum protein. Albumin plays important physiological roles including maintenance of colloid osmotic pressure and binding of key substances such as long-chain fatty acids bile acids bilirubin hematin calcium and magnesium. It has antioxidant and anticoagulant effects acts as a carrier for nutritional factors and drugs and is an effective plasma pH buffer. Serum albumin is a reliable prognostic indicator for morbidity and mortality liver disease nephritic syndrome malnutrition and protein-losing enteropathies. High levels are associated with dehydration.
Catalog No. 50-290-4889 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1241KT
Albumin is the most abundant plasma protein in humans. It accounts for 60% of the total serum protein. Albumin plays important physiological roles including maintenance of colloid osmotic pressure and binding of key substances such as long-chain fatty acids bile acids bilirubin hematin calcium and magnesium. It has antioxidant and anticoagulant effects acts as a carrier for nutritional factors and drugs and is an effective plasma pH buffer. Serum albumin is a reliable prognostic indicator for morbidity and mortality liver disease nephritic syndrome malnutrition and protein-losing enteropathies. High levels are associated with dehydration.