Learn More
New England Biolabs, Inc. BcgI – 250 units
SDP
Catalog No. 50812221
Encompass
A restriction endonuclease that recognizes the sequence _NN^NNNNNNNNNNCGANNNNNNTGCNNNNNNNNNN_NN^.
Catalog No. 50-812-221 Supplier New England Biolabs, Inc. Supplier No. R0545S
A restriction endonuclease that recognizes the sequence _NN^NNNNNNNNNNCGANNNNNNTGCNNNNNNNNNN_NN^.