Learn More
Selleck Chemical LLC BCH-5mg
SDP
Catalog No. 502389758
Encompass_Preferred
BCH (2-Aminobicyclo-(2,2,1)-heptane-2-carboxylic acid, LAT1-IN-1) is a selective and competitive inhibitor of system L amino acid transporter 1 (LAT1). BCH (LAT1-IN-1) induces apoptosis in cancer cells.
Catalog No. 50-238-9758 Supplier Selleck Chemical LLC Supplier No. S68945MG
BCH (2-Aminobicyclo-(2,2,1)-heptane-2-carboxylic acid, LAT1-IN-1) is a selective and competitive inhibitor of system L amino acid transporter 1 (LAT1). BCH (LAT1-IN-1) induces apoptosis in cancer cells.