Learn More
Medchemexpress LLC HY-21972 5mg Medchemexpress, BCI-121 CAS:432529-82-3 Purity:>98%
SDP
Catalog No. 502082218
Encompass_Preferred
Medchemexpress, HY-21972 5mg BCI-121 CAS:432529-82-3 BCI-121 is a SMYD3 inhibitor that impairs the proliferation of cancer cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2218 Supplier Medchemexpress LLC Supplier No. HY219725MG
Medchemexpress, HY-21972 5mg BCI-121 CAS:432529-82-3 BCI-121 is a SMYD3 inhibitor that impairs the proliferation of cancer cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.