Learn More
TARGETMOL CHEMICALS INC BCI-137 10MG
SDP
Catalog No. 502877612
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. BCI-137 is an Argonaute 2 ligand. It acts by targeting the MicroRNA binding domain. Purity 95%
Catalog No. 50-287-7612 Supplier TARGETMOL CHEMICALS INC Supplier No. T2377710MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. BCI-137 is an Argonaute 2 ligand. It acts by targeting the MicroRNA binding domain. Purity 95%