Learn More
Medchemexpress LLC HY-121087 5mg Medchemexpress, BCI-215 CAS:1245792-67-9 Purity:>98%
SDP
Catalog No. 501969617
Encompass_Preferred
Medchemexpress, HY-121087 5mg BCI-215 CAS:1245792-67-9 BCI-215 is a potent and tumor cell-selective dual specificity MAPK phosphatase (DUSP-MKP) inhibitor. BCI-215 has cytotoxicity for tumor cells but not normal cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9617 Supplier Medchemexpress LLC Supplier No. HY1210875MG
Medchemexpress, HY-121087 5mg BCI-215 CAS:1245792-67-9 BCI-215 is a potent and tumor cell-selective dual specificity MAPK phosphatase (DUSP-MKP) inhibitor. BCI-215 has cytotoxicity for tumor cells but not normal cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.