Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical BCI™ 3301 Pulse Oximeter

Catalog No. 19815372
Change view
Click to view available options
Display Type:
LED
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
19-815-372 LED
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-815-372 Supplier Moore Medical Supplier No. 48191

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

reusable adult finger probe, and three C batteries

Provides fast, reliable SpO2 and pulse rate measurements on all patients, neonate to adult

  • Durable, high-performance unit that is both easy to use and completely transportable
  • Designed for simple, accurate monitoring, this unit ensures reliable, consistent oxygenation surveillance
  • Weighs 16.3 oz.
  • Easy to read LED display
  • Simple to use on/off controls
  • Auto shut-off feature turns unit off when finger is removed from probe
  • Quantitative pulse-strength bar shows actual pulse strength and perfusion status
  • Unique memory feature and hard copy documentation available for up to 99 patient SpO2 and pulse-rate readings

Specifications

Product Type 3301 Pulse Oximeter
Display Type LED
Weight (English) 16.3 oz.
Weight (Metric) 0.46 kg
For Use With (Application) To read display and accurately indicates SpO2 and pulse rate for patients from neonate to adult
Includes Reusable adult finger probe and three C batteries
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.