Learn More
Medchemexpress LLC HY-115502A 1mg Medchemexpress, BCI (hydrochloride) CAS:95130-23-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY115502A1MG
Encompass_Preferred
Medchemexpress, HY-115502A 1mg BCI (HCl) CAS:95130-23-7 BCI hydrochloride ((E)-BCI hydrochloride) is an allosteric inhibitor of dual specificity phosphatase (DUSP). BCI hydrochloride specifically inhibits DUSP6 and DUSP1 with EC50s of 13.3 and 8.0 μM in cells, respectively. BCI hydrochloride does not inhibit DUSP5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9389