Learn More
Medchemexpress LLC BCI hydrochloride 10mg | 95130-23-7 | 353.89 g/mol | C22H24ClNO | 10 MG
SDP
Catalog No. 5000227389
Encompass_Preferred
BCI hydrochloride is a small-molecule inhibitor of dual-specificity phosphatase 6 (DUSP6) used in research on inflammation and oxidative stress. It has demonstrated anti-inflammatory activity, reduces reactive oxygen species production, and modulates pro-inflammatory cytokine expression in activated macrophages.
  • Potent inhibitor of DUSP6 with demonstrated cellular activity.
  • Reduces reactive oxygen species and activates the Nrf2 pathway in macrophages.
  • Inhibits expression of IL-1β and IL-6 in LPS-activated macrophages.
  • High reported purity suitable for research applications.
  • Soluble in DMSO (approx. 15.62 mg/mL) for in vitro use.
  • Provided as a stable solid for storage at -20°C under sealed, dry conditions.
Catalog No. 50-002-27389 Supplier Medchemexpress LLC Supplier No. HY115502A10MG
BCI hydrochloride is a small-molecule inhibitor of dual-specificity phosphatase 6 (DUSP6) used in research on inflammation and oxidative stress. It has demonstrated anti-inflammatory activity, reduces reactive oxygen species production, and modulates pro-inflammatory cytokine expression in activated macrophages.
  • Potent inhibitor of DUSP6 with demonstrated cellular activity.
  • Reduces reactive oxygen species and activates the Nrf2 pathway in macrophages.
  • Inhibits expression of IL-1β and IL-6 in LPS-activated macrophages.
  • High reported purity suitable for research applications.
  • Soluble in DMSO (approx. 15.62 mg/mL) for in vitro use.
  • Provided as a stable solid for storage at -20°C under sealed, dry conditions.