Learn More
Medchemexpress LLC BCI hydrochloride 10mM 1mL | 95130-23-7 | 353.89 | C22H24ClNO | 1 ML
SDP
Catalog No. 5000228807
Encompass_Preferred
BCI hydrochloride is the hydrochloride salt of (E)-BCI, a small-molecule inhibitor of dual specificity phosphatase 6 (DUSP6). It is supplied as a 10 mM solution in DMSO (1 mL) or as solid samples for biochemical and cellular research into DUSP6-related signaling, inflammation, and reactive oxygen species (ROS).
  • Allosteric DUSP6 inhibitor used in biochemical and cellular assays.
  • Exhibits anti-inflammatory activity and reduces ROS production.
  • Available as a 10 mM, 1 mL solution in DMSO or as solid packs (1-500 mg).
  • CAS number 95130-23-7; molecular weight 353.89 g/mol.
  • Light yellow to yellow solid; soluble in DMSO.
Catalog No. 50-002-28807 Supplier Medchemexpress LLC Supplier No. HY115502A10MM 1ML
BCI hydrochloride is the hydrochloride salt of (E)-BCI, a small-molecule inhibitor of dual specificity phosphatase 6 (DUSP6). It is supplied as a 10 mM solution in DMSO (1 mL) or as solid samples for biochemical and cellular research into DUSP6-related signaling, inflammation, and reactive oxygen species (ROS).
  • Allosteric DUSP6 inhibitor used in biochemical and cellular assays.
  • Exhibits anti-inflammatory activity and reduces ROS production.
  • Available as a 10 mM, 1 mL solution in DMSO or as solid packs (1-500 mg).
  • CAS number 95130-23-7; molecular weight 353.89 g/mol.
  • Light yellow to yellow solid; soluble in DMSO.