Learn More
Medchemexpress LLC BCI hydrochloride 5mg | 95130-23-7 | 5 MG
SDP
Catalog No. 5000227365
Encompass_Preferred
BCI hydrochloride is the hydrochloride salt of (E)-BCI, a small-molecule inhibitor of dual-specificity phosphatases DUSP6 and DUSP1. It is supplied as a solid research reagent for biochemical and cell-based studies to modulate MAPK pathway signalling.
  • Selective DUSP6 and DUSP1 inhibitor useful for MAPK pathway studies.
  • High chemical purity (99.93% as listed by the supplier).
  • Solid form, suitable for storage and dissolution in common solvents.
  • Supplied in a 5 MG quantity for small-scale laboratory research.
  • Identified by CAS 95130-23-7 for unambiguous compound reference.
Catalog No. 50-002-27365 Supplier Medchemexpress LLC Supplier No. HY115502A5MG
BCI hydrochloride is the hydrochloride salt of (E)-BCI, a small-molecule inhibitor of dual-specificity phosphatases DUSP6 and DUSP1. It is supplied as a solid research reagent for biochemical and cell-based studies to modulate MAPK pathway signalling.
  • Selective DUSP6 and DUSP1 inhibitor useful for MAPK pathway studies.
  • High chemical purity (99.93% as listed by the supplier).
  • Solid form, suitable for storage and dissolution in common solvents.
  • Supplied in a 5 MG quantity for small-scale laboratory research.
  • Identified by CAS 95130-23-7 for unambiguous compound reference.