Learn More
Abcam BCIP disodium salt, histochemical alkaline phosphatase substrate, 500MG
Catalog No. 502704822
Encompass_Preferred
MW 370.43 g/mol. Chromogenic substrate for alkaline phosphatase. Often used in combination with nitro blue tetrazolium chloride (NBT) to amplify the sensitivity of staining.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4822 Supplier Abcam Supplier No. AB146226500MG
MW 370.43 g/mol. Chromogenic substrate for alkaline phosphatase. Often used in combination with nitro blue tetrazolium chloride (NBT) to amplify the sensitivity of staining.
The product is subject to the following: Abcam Restricted Use Statement