Learn More
Seracare Life Sciences Inc BCIP/NBT 1-C AP SUBSTRATE
Supplier:  Seracare Life Sciences Inc 54200036
Encompass_Preferred
Effective chromogenic substrate for use in blotting, it provides crisp bands of intense purple precipitate with minimal background staining. Designed for use with alkaline phosphatase-labeled conjugates. Recommended for blotting and ELIspot Assays. 1-Component format of BCIP/NBT (5-bromo-4-chloro-3-indoyl-phosphate/nitroblue tetrazolium), no reagent preparation, hence, more convenient.
Catalog No. 50-674-34