Learn More
Seracare Life Sciences Inc BCIP/NBT 1-C AP SUBSTRATE
Catalog No. 5067433
Encompass_Preferred
Effective chromogenic substrate for use in blotting, it provides crisp bands of intense purple precipitate with minimal background staining. Designed for use with alkaline phosphatase-labeled conjugates. Recommended for blotting and ELIspot Assays. 1-Component format of BCIP/NBT (5-bromo-4-chloro-3-indoyl-phosphate/nitroblue tetrazolium), no reagent preparation, hence, more convenient.
This item is not returnable. View return policy
Catalog No. 50-674-33 Supplier Seracare Life Sciences Inc Supplier No. 54200033
Effective chromogenic substrate for use in blotting, it provides crisp bands of intense purple precipitate with minimal background staining. Designed for use with alkaline phosphatase-labeled conjugates. Recommended for blotting and ELIspot Assays. 1-Component format of BCIP/NBT (5-bromo-4-chloro-3-indoyl-phosphate/nitroblue tetrazolium), no reagent preparation, hence, more convenient.