Learn More
Seracare Life Sciences Inc BCIP/NBT AP SUBSTRATE SYSTEM
Catalog No. 5067432
Encompass_Preferred
Effective chromogenic substrate for use in blotting, it provides crisp bands of intense purple precipitate with minimal background staining. Designed for use with alkaline phosphatase-labeled conjugates. 3-Component format of BCIP/NBT (5-bromo-4-chloro-3-indoyl-phosphate/nitroblue tetrazolium), simply mix and use.
This item is not returnable. View return policy
Catalog No. 50-674-32 Supplier Seracare Life Sciences Inc Supplier No. 54200030
Effective chromogenic substrate for use in blotting, it provides crisp bands of intense purple precipitate with minimal background staining. Designed for use with alkaline phosphatase-labeled conjugates. 3-Component format of BCIP/NBT (5-bromo-4-chloro-3-indoyl-phosphate/nitroblue tetrazolium), simply mix and use.