Learn More
Surmodics Inc BCIP/NBT Purple One Component AP Membrane Substrate, 1L
SDP
Catalog No. NC0927674
Encompass_Preferred
The substrate is a ready to use, one-component formulation of 5-bromo-4-chloro-3-indolylphosphate and nitroblue tetrazolium for use in immunoblotting or other membrane applications.
Catalog No. NC0927674 Supplier Surmodics Inc Supplier No. BCID100001
The substrate is a ready to use, one-component formulation of 5-bromo-4-chloro-3-indolylphosphate and nitroblue tetrazolium for use in immunoblotting or other membrane applications.