Learn More
Crescent Chemical Co Inc BCIP/NBT READY-TO-USE
SDP
Catalog No. 50247374
Encompass_Preferred
BCIP/NBT ready-to-use substrate for immunohistochemistry and blotting Single component substrate solution for detection of alkaline phosphatase in immunohistochemical, in situ hybridization and blotting procedures. A very fine blue-purple precipitate will be localized at sites of
Catalog No. 50-247-374 Supplier Crescent Chemical Co Inc Supplier No. SE15246.01
BCIP/NBT ready-to-use substrate for immunohistochemistry and blotting Single component substrate solution for detection of alkaline phosphatase in immunohistochemical, in situ hybridization and blotting procedures. A very fine blue-purple precipitate will be localized at sites of