Learn More
Vector Laboratories BCIP/NBT Alkaline Phosphatase (AP) Substrate Kit 200 ml
SDP
Catalog No. NC9664831
Encompass
BCIP/NBT Alkaline Phosphatase (AP) Substrate Kit, 5-bromo-4-chloro-3-indolyl phosphate/nitroblue tetrazolium, forms an indigo, heat-stable reaction product that can be dehydrated, cleared, and permanently mounted. This substrate can also be used for developing blots or for in situ hybridization. With 20-30 minute development times, the sensitivity of this substrate is equivalent to the other alkaline phosphatase substrates. However, BCIP/NBT will continue to develop from several hours to overnight making it one of the most sensitive chromogenic substrates. The stock solutions in this kit are provided in convenient dropper bottles, in sufficient volume for 200 ml working solution.
Catalog No. NC9664831 Supplier Vector Laboratories Supplier No. SK5400
BCIP/NBT Alkaline Phosphatase (AP) Substrate Kit, 5-bromo-4-chloro-3-indolyl phosphate/nitroblue tetrazolium, forms an indigo, heat-stable reaction product that can be dehydrated, cleared, and permanently mounted. This substrate can also be used for developing blots or for in situ hybridization. With 20-30 minute development times, the sensitivity of this substrate is equivalent to the other alkaline phosphatase substrates. However, BCIP/NBT will continue to develop from several hours to overnight making it one of the most sensitive chromogenic substrates. The stock solutions in this kit are provided in convenient dropper bottles, in sufficient volume for 200 ml working solution.