Learn More
Alkali Scientific BCIP, P Toluidine Salt [5 Bromo 4 Chloro 3 Indolyl Phospate P Toluidine Salt], 1 Gram
SDP
Catalog No. 5000661099
Encompass_Preferred
BCIP (5-Bromo 4-Chloro 3-Indolyl Phosphate P-Toluidine Salt) is a chromogenic substrate used for detecting alkaline phosphatase activity in molecular biology applications. This reagent produces a colored product upon cleavage by alkaline phosphatase, which can be used to detect enzyme activity in assays such as Southern blots, Northern blots, and ELISAs. The 1g bottle is sufficient for numerous experiments and is widely used in gene expression studies, immunohistochemistry, and other techniques where enzyme activity needs to be visualized. BCIP is a reliable and effective tool for researchers studying phosphatase activity and enzyme-linked assays, providing clear and visible results in biochemical experiments.
Catalog No. 50-006-61099 Supplier Alkali Scientific Supplier No. BCI001CP
BCIP (5-Bromo 4-Chloro 3-Indolyl Phosphate P-Toluidine Salt) is a chromogenic substrate used for detecting alkaline phosphatase activity in molecular biology applications. This reagent produces a colored product upon cleavage by alkaline phosphatase, which can be used to detect enzyme activity in assays such as Southern blots, Northern blots, and ELISAs. The 1g bottle is sufficient for numerous experiments and is widely used in gene expression studies, immunohistochemistry, and other techniques where enzyme activity needs to be visualized. BCIP is a reliable and effective tool for researchers studying phosphatase activity and enzyme-linked assays, providing clear and visible results in biochemical experiments.