Learn More
Alkali Scientific BCIP, P Toluidine Salt [5 Bromo 4 Chloro 3 Indolyl Phospate P Toluidine Salt], 500 Milligrams
SDP
Catalog No. 5000661078
Encompass_Preferred
Indolyl Phosphate Toluidine Salt (500 mg) is a chromogenic substrate used in enzyme detection assays, particularly in the study of phosphatases. Upon enzymatic hydrolysis, the substrate releases a color change, allowing for visual or quantitative measurement of enzyme activity. This reagent is frequently used in clinical diagnostics, enzyme research, and protein assays. The 500 mg formulation is suitable for medium-scale assays and is ideal for studying enzyme kinetics and enzyme inhibition. It provides researchers with an effective and sensitive method for detecting enzymatic activity in various biological systems, including clinical and pharmaceutical applications.
Catalog No. 50-006-61078 Supplier Alkali Scientific Supplier No. BCI004CP
Indolyl Phosphate Toluidine Salt (500 mg) is a chromogenic substrate used in enzyme detection assays, particularly in the study of phosphatases. Upon enzymatic hydrolysis, the substrate releases a color change, allowing for visual or quantitative measurement of enzyme activity. This reagent is frequently used in clinical diagnostics, enzyme research, and protein assays. The 500 mg formulation is suitable for medium-scale assays and is ideal for studying enzyme kinetics and enzyme inhibition. It provides researchers with an effective and sensitive method for detecting enzymatic activity in various biological systems, including clinical and pharmaceutical applications.