Learn More
AdipoGen BCIP p-toluidine salt
SDP
Supplier:  AdipoGen CDXB0024M100
Encompass_Preferred
Chemical. CAS 6578-06-9. Formula C8H6BrClNO4P . C7H9N. MW 433.62. BCIP is a colorimetric substrate for alkaline phosphatase. Upon enzymatic cleavage by alkaline phosphatase, an indoxyl intermediate is released and subsequently oxidized to produce water-insoluble indigo. It can be used to detect alkaline phosphatase activity by immunohistochemistry IHC or Western blot WB.
Catalog No. 50-256-1706